Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ZL55 cell line human

12050301, human pleural cavity, Epitheloid/mesenchymal

Synonym(s):

ZL 55, ZL-55

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

ZL55 cell line human, 12050301

biological source

human pleural cavity

packaging

tube of 5 μg 12050301-DNA-5UG
pkg of vial of cells 12050301-1VL

growth mode

adherent

morphology

Epitheloid/mesenchymal

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Cell Line Description

ZL55 was derived from an epithelial tumor of the pleural cavity of a 52-year old male, prior to treatment, with a known history of exposure to asbestos.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 9,11
D13S317: 8
D16S539: 12
D5S818: 10,13
D7S820: 7,11
THO1: 6
TPOX: 8
vWA: 15,19

Culture Medium

DMEM:HAMS F12 (1:1) + 2mM Glutamine + 15% Fetal Bovine Serum (FBS)

Subculture Routine

Split subconfluent cultures (70-80%) 1:4 to 1:10 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Suggested seeding density 1-4 x 10,000 cells/cm².

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service