Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

YTH 913.12

11060810

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Rat Hybridoma

ImmunoDon

DA rat spleen

Myeloma

Y3.AG.1.2.3

Reactivity

Human CD28

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Monoclonal antibody, mouse CD45

Cell Line Description

Rat hybridoma producing antibody against human CD28. Cambridge Enterprise Reference number AB113:www.enterprise.cam.ac.uk.

Immunogen

Activated human T cells

Culture Medium

IDMEM (I3390) + 2 mM L-Glutamine (G7513) + 5% FBS / FCS (F2442). Where the cells are grown under static conditions, 0.02% EDTA, Trypsin (T4424) or mechanical agitation maybe required to dislodge them from the plastic.

Subculture Routine

Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37 °C. Saturation density 9 x 105/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service