Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HeLa9903

11033105, human cervix, Epithelial

Synonym(s):

HeLa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

HeLa9903, 11033105, from human cervix

biological source

human cervix

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

HeLa, genetic modification, the hER alpha expression construct, Oestrogen-Responsive Element

Cell Line Description

HeLa9903 is a HeLa cell line stably transfected with two constructs: the hER alpha expression construct; a firefly luciferase reporter construct bearing Oestrogen-Responsive Element (ERE) driven by a mouse metallothionein (MT) promoter TATA element. HeLa9903 can be used to test for evaluation of chemicals based on OECD Test Guideline: The stably transformed human oestrogen receptor-alpha transcriptional; activation assay for detection of oestrogenic activity of chemicals.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 9,10
D16S539: 9,10
D5S818: 11,12
D7S820: 8,12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

DMEM + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS) + 16μg/ml Blasticidin S HCL + 800μg/ml G418.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 , i.e., seeding at 2 x 104 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

US7537904B1 N Ooe & Matsunaga H 2009.Cell for measuring the ability to control the activity of a ligand-responsive transcription control factor.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service