Skip to Content
MilliporeSigma
All Photos(1)

Documents

4-11

11033103, human cervix, Epithelial

Synonym(s):

HeLa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

4-11, 11033103

biological source

human cervix

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

HeLa, genetic modification, human androgen receptor, firefly luciferase reporter gene, MMTV promoter

Cell Line Description

4-11 is a HeLa cell line stably transfected with two constructs: the human androgen receptor (hAR) expression construct; a firefly luciferase reporter gene under the control of the MMTV promoter. 4-11 can be used to detect androgenic activity of chemicals.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 9,10
D16S539: 9,10
D5S818: 11,12
D7S820: 8,12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

DMEM (D6546) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442) + 16 μg/ml Blasticidin S HCL + 800 μg/ml G418.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10, i.e. seeding at 2 x 104 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

US7537904B1 N Ooe & Matsunaga H 2009.Cell for measuring the ability to control the activity of a ligand-responsive transcription control factor.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service