Skip to Content
MilliporeSigma
All Photos(1)

Documents

MM138 Cell Line human

10092321, human skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MM138 Cell Line human, 10092321

biological source

human skin

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human melanoma; from metastatic site - lymph node

Cell Line Description

Metastatic Melanoma (metastatic site: lymph node). Cell Bank Australia reference number: CBA1345.

Culture Medium

RPMI 1640 (with 2 mM L-Glutamine + 25 mM HEPES) + 10% fetal bovine serum (FBS).

Subculture Routine

Optimal split ratio 1:4, MM138 has a relatively slow doubling time of around 60 hours. It does not become confluent and will detach after 6-7 days. If split too hard the doubling time is greatly increased. (Seeding density 0.8 x104 cells/cm2). Harvest the cells using 0.05% Trypsin/EDTA at 37 °C for 5 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service