Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

utr-1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Human x Mouse Hybridoma

packaging

tube of 5 μg 10080904-DNA-5UG
pkg of vial of cells 10080904-1VL

ImmunoDon

Mouse (Mus musculus) spleen

Myeloma

PAI

Reactivity

Human CD120b (TNF receptor p75)

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Application

Sandwich ELISA for soluble p75 TNF-R. No competition with TNF.

Culture Medium

Iscoves Modified Dulbeccos Medium (IMDM) + 2 mM glutamine + 25 U/ml Interleukin-6 (IL-6) + 5-10% Fetal Bovine Serum (FBS). Where the cells are grown under static conditions, 0.02% EDTA may be required to dislodge them from the plastic.

Subculture Routine

Seed cultures between 1-3 x 105 cells/ml; Saturation density 8-10 x 105 5% CO2; 37 °C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service