Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.4E7

10070102, human pancreas (islets), Epitheloid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

1.4E7, 10070102, from human pancreas(islets), NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

human pancreas (islets)

packaging

tube of 5 μg 10070102-DNA-5UG
pkg of vial of cells 10070102-1VL

growth mode

Not specified

karyotype

Modal chromosome number 67-72

morphology

Epitheloid

products

1.4E7 cells have been shown to express insulin, glucokinase and IAPP by immunocytochemistry. These cells express the GLUT1 glucose transporter.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Description

1.4E7 is a hybrid cell line formed by the electrofusion of a primary culture of human pancreatic islets with PANC-1, a human pancreatic ductal carcinoma cell line (ECACC catalogue number 87092802). 1.4E7 has been shown to be tumourigenic when transplanted into a SCID mouse host. The cell line has applications in the study of pancreatic cell biology. 1.4E7 cells provide a method of producing pure insulin secreting cells when stimulated. This is an alternative to the use of primary tissue in cell transplantation therapies for type 1 diabetes. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 10,12
D13S317: 11
D16S539: 11
D5S818: 11,13
D7S820: 8
THO1: 7,8
TPOX: 8,11
vWA: 15

Culture Medium

RPMI-1640 + 2 mM Glutamine + 10% FCS / FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 , i.e., seeding at 2-4 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Population doubling approx 26hrs. At confluence 105 cells/cm2 can be expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

Flatt P & McClenaghan N (2004) Insulin producing cell-line US patent number 20040005702 (A1). Also WO0210346 (A2).

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service