UM-RC-3
8090512, human kidney (renal gland), Polygonal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
Product Name
UM-RC-3, 08090512
biological source
human kidney (renal gland)
growth mode
Adherent
karyotype
Median number of chromosomes is 55
morphology
Polygonal
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human renal carcinoma
Cell Line Description
UM-RC-3 is a continuous cell line established from a primary renal adenocarcinoma. It exhibits a loss of contact inhibition and produces tumours in the nude mouse model. The patient had metastatic disease at the time of their nephrectomy.
Application
Study of tumour associated antigens.
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 9,12
D16S539: 9
D5S818: 9,12
D7S820: 9
THO1: 9.3
TPOX: 8
vWA: 16,18
CSF1PO: 12
D13S317: 9,12
D16S539: 9
D5S818: 9,12
D7S820: 9
THO1: 9.3
TPOX: 8
vWA: 16,18
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Doubling time approximately 29 hours.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service