Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A9(Hamprecht)

8062526, mouse connective tissue, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

A9(Hamprecht), 08062526

biological source

mouse connective tissue

growth mode

Adherent

karyotype

Modal No of chromosomes is 54

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Fibroblast (L cells)

Cell Line Description

A 6-thioguanine resistant mutant of mouse L cell fibroblasts (Mouse strain C3H/An). A9 lacks guanylic acid-inosinic acid pyrophosphorylase and was isolated by virtue of resistance to 6-thioguanine (3 μg/ml). ECACC has two catalogue entries for the A9 cell line which differ in that they were deposited by different groups: A9 (Sigma Catalogue number 85011426) and A9(Hamprecht) (Sigma Catalogue number 08062526). Several other cell lines in the ECACC catalogue have been derived from A9(Hamprecht).

Application

Used in the generation of cell hybrids.

Culture Medium

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + 0.1 mM 6-thioguanine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service