Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1411H

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 6011805, human embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human embryo

packaging

tube of 5 μg 06011805-DNA-5UG
pkg of vial of cells 06011805-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human yolk sac carcinoma

Cell Line Description

A non seminomatous germ cell carcinoma derived cell line. A yolk sac carcinoma cell line derived from a testicular teratomacarcinoma. This cell line is tumourigenic in nude mice.

Culture Medium

DMEM (High Glucose) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS). No sodium pyruvate.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 10% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service