Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

577MF

6011802, human forehead teratoma, Teratoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

577MF, 06011802

biological source

human forehead teratoma

growth mode

Adherent

karyotype

Not specified

morphology

Teratoma

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human forehead metastasis of malignant teratoma

Cell Line Description

An undifferentiated carcinoma cell line established from a forehead metastasis of a malignant teratoma. This is not an embryonal carcinoma cell line. This cell line is tumourigenic in nude mice.

Culture Medium

DMEM (High Glucose) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin or trypsin/EDTA; 10% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service