Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9994

Sigma-Aldrich

CAMK2β, active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAM2, CAMKB, MG C29528

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

4820-6521 nmol/min·mg

mol wt

~58 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CAMK2B(816)

Biochem/physiol Actions

CAMK2β belongs to the serine/threonine protein kinase family and to the type II multifunctional Ca2+/calmodulin-dependent protein kinase subfamily. CAMK2β showed wide tissue and cell distribution, and one of CAMK2β variant predominated in adult brain. The ratio of CAMK2α and CAMK2β protein levels were inversely related during activity in hippocampal neurons. CAMK2β is a prominent kinase in the central nervous system and may function in long-term potentiation and neurotransmitter release.

Physical form

Supplied in 50mM NaPhosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1mM PMSF, 0.2 mM DTT, 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tara C Thiagarajan et al.
Neuron, 36(6), 1103-1114 (2002-12-24)
We show that alpha and betaCaMKII are inversely regulated by activity in hippocampal neurons in culture: the alpha/beta ratio shifts toward alpha during increased activity and beta during decreased activity. The swing in ratio is approximately 5-fold and may help
R M Tombes et al.
Biochimica et biophysica acta, 1355(3), 281-292 (1997-03-01)
CaMK-II (the (type II) multifunctional Ca2+/CaM-dependent protein kinase) has been implicated in diverse neuronal and non-neuronal functions, including cell growth control. CaMKII expression was evaluated in a variety of human tumor cell lines using RT-PCR (reverse transcriptase coupled polymerase chain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service