Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9984

Sigma-Aldrich

Z-Gly-Leu-Phe-chloromethyl ketone

≥90% (TLC), solid

Synonym(s):

Z-GLF-CMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N3O5Cl
CAS Number:
Molecular Weight:
502.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥90% (TLC)

form

solid

solubility

DMSO: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)CNC(=O)OCc1ccccc1)C(=O)N[C@@H](Cc2ccccc2)C(=O)CCl

InChI

1S/C26H32ClN3O5/c1-18(2)13-22(25(33)30-21(23(31)15-27)14-19-9-5-3-6-10-19)29-24(32)16-28-26(34)35-17-20-11-7-4-8-12-20/h3-12,18,21-22H,13-17H2,1-2H3,(H,28,34)(H,29,32)(H,30,33)/t21-,22-/m0/s1

InChI key

BRGPJZPHCVTNNI-VXKWHMMOSA-N

Amino Acid Sequence

Z-Gly-Leu-Phe-CMK

Biochem/physiol Actions

Cathepsin G inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substrate binding site in bovine chymotrypsin A-gamma. A crystallographic study using peptide chloromethyl ketones as site-specific inhibitors.
D M Segal et al.
Biochemistry, 10(20), 3728-3738 (1971-09-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service