Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9794

Sigma-Aldrich

(±)-Z-Phosphonoglycine trimethyl ester

Synonym(s):

(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl) acetate, (±)-Trimethyl Z-phosphonoglycinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
331.26
Beilstein:
5764460
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

mp

77-80 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)C(NC(=O)OCc1ccccc1)P(=O)(OC)OC

InChI

1S/C13H18NO7P/c1-18-12(15)11(22(17,19-2)20-3)14-13(16)21-9-10-7-5-4-6-8-10/h4-8,11H,9H2,1-3H3,(H,14,16)

InChI key

GSYSFVSGPABNNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes; the dehydroamino acids can be stereoselectively reduced to amino acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service