Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9504

Sigma-Aldrich

Coenzyme Q6

Synonym(s):

2,3-Dimethoxy-5-methyl-6-(farnesylfarnesyl)-1,4-benzoquinone, Ubiquinone-30

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H58O4
CAS Number:
Molecular Weight:
590.88
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

biological source

Saccharomyces cerevisiae

storage temp.

−20°C

SMILES string

COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C)=C(C)C1=O

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Ozeir et al.
Chemistry & biology, 18(9), 1134-1142 (2011-09-29)
Coenzyme Q (Q), an essential component of eukaryotic cells, is synthesized by several enzymes from the precursor 4-hydroxybenzoic acid. Mutations in six of the Q biosynthesis genes cause diseases that can sometimes be ameliorated by oral Q supplementation. We establish here
Mario H Barros et al.
The Journal of biological chemistry, 280(52), 42627-42635 (2005-10-19)
Deletion of the Saccharomyces cerevisiae gene YOL008W, here referred to as COQ10, elicits a respiratory defect as a result of the inability of the mutant to oxidize NADH and succinate. Both activities are restored by exogenous coenzyme Q2. Respiration is
Beth Marbois et al.
The Journal of biological chemistry, 280(21), 20231-20238 (2005-03-29)
Coenzyme Q (Q) is a redox active lipid essential for aerobic respiration in eukaryotes. In Saccharomyces cerevisiae at least eight mitochondrial polypeptides, designated Coq1-Coq8, are required for Q biosynthesis. Here we present physical evidence for a coenzyme Q-biosynthetic polypeptide complex
Sergio Padilla-López et al.
Biochimica et biophysica acta, 1788(6), 1238-1248 (2009-04-07)
Coenzyme Q is an isoprenylated benzoquinone lipid that functions in respiratory electron transport and as a lipid antioxidant. Dietary supplementation with Q is increasingly used as a therapeutic for treatment of mitochondrial and neurodegenerative diseases, yet little is known regarding
Alejandro Martín-Montalvo et al.
The Biochemical journal, 440(1), 107-114 (2011-08-05)
CoQ(6) (coenzyme Q(6)) biosynthesis in yeast is a well-regulated process that requires the final conversion of the late intermediate DMQ(6) (demethoxy-CoQ(6)) into CoQ(6) in order to support respiratory metabolism in yeast. The gene CAT5/COQ7 encodes the Cat5/Coq7 protein that catalyses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service