Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9419

Sigma-Aldrich

4-Chloro-D-phenylalanine

Synonym(s):

D-PCP, D-PCPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10ClNO2
CAS Number:
Molecular Weight:
199.63
Beilstein:
2416151
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

technique(s)

ligand binding assay: suitable

storage temp.

−20°C

SMILES string

N[C@H](Cc1ccc(Cl)cc1)C(O)=O

InChI

1S/C9H10ClNO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m1/s1

InChI key

NIGWMJHCCYYCSF-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

4-Chloro-D-phenylalanine (D-PCP, D-Phe(4Cl)) is a component of the synthetic decapeptide SB-4-Chloro-D-phenylalanine. (D-PCP, D-Phe(4Cl)) is a component of the synthetic peptides SB-75/cetrorelix, a Gonadotropin Releasing Hormone (GnRH) antagonist, and of the VIP receptor antagonist (4Cl-D-Phe(6),Leu(17))-VIP.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kordasti et al.
Gut, 53(7), 952-957 (2004-06-15)
The mechanisms underlying intestinal secretion in rotavirus diarrhoea remain to be established. We previously reported that rotavirus evokes intestinal fluid and electrolyte secretion by activation of the enteric nervous system. We now report that antagonists for the 5-hydroxytryptamine 3 receptor
J Pinski et al.
International journal of peptide and protein research, 45(5), 410-417 (1995-05-01)
The objective of this study was to examine the in vivo and in vitro gonadotropin-inhibiting potencies, edematogenic activities and the receptor binding affinities of the D-Cit6, D,L-Cit6 and L-Cit6 forms of the LH-RH antagonist Cetrorelix (SB-75) [Ac-D-Nal(2)1,D-Phe(4Cl)2,D-Pal(3)3,D-Cit6,D-Ala10]LH- RH. In order
T Yano et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(15), 7090-7094 (1994-07-19)
Female athymic nude mice bearing xenografts of OV-1063 human epithelial ovarian cancer cell line were treated with potent luteinizing hormone (LH)-releasing hormone (LH-RH) antagonist SB-75 (Cetrorelix; [Ac-D-Nal(2)1, D-Phe(4 CI)2, D-Pal(3)3, D-Cit6, D-Ala10]LH-RH in which Ac-D-Nal(2) = N-acetyl-3-(2-naphthyl)-D-alanine, D-Phe(4CI) = 4-chloro-D-phenylalanine
J Horvath et al.
International journal of oncology, 6(5), 969-975 (1995-05-01)
The effects of luteinizing hormone-releasing hormone (LH-RH), and LH-RH antagonist Cetrorelix, (SB-75, [Ac-D-Nal(2)(1),D-Phe(4-Cl)(2),D-Pal(3)(3),D-Cit(6),D-Ala(10)]LH-RH) on cell growth and the production of hCG and cAMP in JAR human choriocarcinoma cells were examined in vitro. Both LH-RH and its antagonist SE-75, at 1
Takeharu Niioka et al.
Archives of oral biology, 54(10), 909-916 (2009-08-04)
The parasympathetic vasodilatory fibres are known to innervate vessels in a rat masseter muscle via both cholinergic and non-cholinergic mechanisms. However, the non-cholinergic mechanisms are still unclear. Recently, vasoactive intestinal polypeptide (VIP) was convincingly shown to be involved in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service