Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9392

Sigma-Aldrich

(±)-Coniine

≥98%, liquid

Synonym(s):

2-Propylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N
CAS Number:
Molecular Weight:
127.23
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98%

form

liquid

color

, clear colorless to yellow-green

density

0.85 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCC1CCCCN1

InChI

1S/C8H17N/c1-2-5-8-6-3-4-7-9-8/h8-9H,2-7H2,1H3

InChI key

NDNUANOUGZGEPO-UHFFFAOYSA-N

Application

Coniine, a neurotoxic alkaloid from hemlock, is used in comparative analysis versus other alkaloids that act at the level of the nicotinic acid/nicotinic acetylcholine (nAChR) receptors.

Other Notes

Toxic principle of poison hemlock (racemic modification).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

119.3 °F - closed cup

Flash Point(C)

48.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benedict T Green et al.
Neurotoxicology and teratology, 32(3), 383-390 (2010-02-02)
Teratogenic alkaloids are found in many species of plants including Conium maculatum L., Nicotiana glauca, Nicotiana tabaccum, and multiple Lupinus spp. Fetal musculoskeletal defects produced by alkaloids from these plants include arthrogyropisis, scoliosis, torticollis, kyposis, lordosis, and cleft palate. A
Anthony J Burke et al.
Organic & biomolecular chemistry, 2(9), 1387-1394 (2004-04-24)
Conjugate addition of lithium (S)-N-benzyl-N-alpha-methylbenzylamide to a range of alpha, beta-unsaturated Weinreb amides proceeds with high levels of diastereoselectivity (>95% de). The beta-amino Weinreb amide products may be transformed into beta-amino ketones via reactions with Grignard reagents, while treatment with
Patrick L West et al.
Pediatric emergency care, 25(11), 761-763 (2009-11-17)
The ingestion of poison hemlock, or Conium maculatum, is described in a 2-year-old boy. He had the onset of abdominal pain and weakness after being fed C. maculatum picked by his sister from the roadside 2 hours earlier. He had
Leo J Schep et al.
Clinical toxicology (Philadelphia, Pa.), 47(8), 771-781 (2009-09-26)
A wide range of plants contain nicotinic and nicotinic-like alkaloids. Of this diverse group, those that have been reported to cause human poisoning appear to have similar mechanisms of toxicity and presenting patients therefore have comparable toxidromes. This review describes
T Blitzke et al.
Phytochemistry, 55(8), 979-982 (2001-01-05)
Phytochemical investigations of Aloe sabaea afforded a new chlorinated amide, N-4'-chlorobutylbutyramide, and the toxic piperidine alkaloids coniine, gamma-coniceine and the quarternary N,N-dimethylconiine. This is the first report of the occurrence of a chlorinated compound in the Aloeaceae family.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service