Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8734

Sigma-Aldrich

Z-Leu-Glu(OMe)-Thr-Asp(OMe)-fluoromethyl ketone

solid

Synonym(s):

Z-LETD-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H44N4O11F
Molecular Weight:
655.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥90% (TLC)

form

solid

storage temp.

−20°C

SMILES string

COC(=O)CC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)OCc1ccccc1)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CC(=O)OC)C(=O)CF

InChI

1S/C30H43FN4O11/c1-17(2)13-22(34-30(43)46-16-19-9-7-6-8-10-19)28(41)32-20(11-12-24(38)44-4)27(40)35-26(18(3)36)29(42)33-21(23(37)15-31)14-25(39)45-5/h6-10,17-18,20-22,26,36H,11-16H2,1-5H3,(H,32,41)(H,33,42)(H,34,43)(H,35,40)/t18-,20+,21+,22+,26+/m1/s1

InChI key

ARECPHKVCIQJRJ-XGRWUTGMSA-N

Amino Acid Sequence

Z-Leu-Glu(OMe)-Thr-Asp(OMe)-FMK

Application

Cell permeable caspase-8 inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guang-quan Li et al.
PloS one, 7(11), e48994-e48994 (2012-11-16)
Dicycloplatin, a new supramolecular platinum-based antitumor drug, has been approved by the State Food and Administration (SFDA) of China. In this study, we investigated the anticancer activity of dicycloplatin in cancer cells and signaling pathways involved in dicycloplatin-induced apoptosis. Dicycloplatin
Xiao-Fan Huang et al.
Molecules (Basel, Switzerland), 25(23) (2020-12-04)
The purpose of the study was to elucidate the anti-hepatoma effects and mechanisms of Pogostemon cablin essential oils (PPa extract) in vitro and in vivo. PPa extract exhibited an inhibitory effect on hepatocellular carcinoma (HCC) cells and was less cytotoxic
Rajeev Kumar et al.
Toxicology and applied pharmacology, 283(3), 187-197 (2015-02-07)
Dietary consumption of phytoestrogens like genistein has been linked with lower incidence of prostate cancer. The estradiol-like benzopyran core of genistein confers estrogen receptor-β (ER-β) selectivity that imparts weak anti-proliferative activity against prostate cancer cells. DL-2-[4-(2-piperidinoethoxy)phenyl]-3-phenyl-2H-1-benzopyran (BP), a SERM designed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service