Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8634

Sigma-Aldrich

Hexyl acetate

Synonym(s):

Capryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)5CH3
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747138
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.409 (lit.)

bp

168-170 °C (lit.)

mp

−80 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCOC(C)=O

InChI

1S/C8H16O2/c1-3-4-5-6-7-10-8(2)9/h3-7H2,1-2H3

InChI key

AOGQPLXWSUTHQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Wu et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(12), 1855-1863 (2015-09-12)
The multispecific organic anion drug transporters OAT6 (SLC22A20) and OAT1 (SLC22A6) are expressed in nasal epithelial cells and both can bind odorants. Sequence analysis of OAT6 revealed an evolutionarily conserved 79-amino acid (AA) fragment present not only in OAT6 but
Zhilin Zhang et al.
Journal of insect science (Online), 14 (2014-12-21)
The morphological structure of the metathoracic scent glands (MTGs) in Adelphocoris suturalis was observed by utilizing scanning electron microscope (SEM). Also, the secretions of MTGs in male and female were analyzed by using gas chromatography-mass spectrometry (GC-MS). The result showed
Xiaobing Pang
Journal of environmental sciences (China), 32, 196-206 (2015-06-05)
Green leaf volatiles (GLVs) emitted by plants after stress or damage induction are a major part of biogenic volatile organic compounds (BVOCs). Proton transfer reaction time-of-flight mass spectrometry (PTR-TOF-MS) is a high-resolution and sensitive technique for in situ GLV analyses
Kye Chung Park et al.
Journal of insect physiology, 78, 33-46 (2015-05-06)
Phytophagous insects detect volatile compounds produced by host and non-host plants, using species-specific sets of olfactory receptor neurons (ORNs). To investigate the relationship between the range of host plants and the profile of ORNs, single sensillum recordings were carried out
Caroline Mongruel Eleutério Dos Santos et al.
Journal of food science, 80(6), C1170-C1177 (2015-04-30)
The amino acid profile in dessert apple must and its effect on the synthesis of fusel alcohols and esters in cider were established by instrumental analysis. The amino acid profile was performed in nine apple musts. Two apple musts with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service