Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C8289

Sigma-Aldrich

Clebopride maleate salt

~95%

Synonym(s):

4-(4-Amino-5-chloro-2-methoxybenzamido)-1-benzylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClN3O2 · C4H4O4
CAS Number:
Molecular Weight:
489.95
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

Assay

~95%

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.COc1cc(N)c(Cl)cc1C(=O)NC2CCN(CC2)Cc3ccccc3

InChI

1S/C20H24ClN3O2.C4H4O4/c1-26-19-12-18(22)17(21)11-16(19)20(25)23-15-7-9-24(10-8-15)13-14-5-3-2-4-6-14;5-3(6)1-2-4(7)8/h2-6,11-12,15H,7-10,13,22H2,1H3,(H,23,25);1-2H,(H,5,6)(H,7,8)/b;2-1+

InChI key

BCVIWCRZYPHHMQ-WLHGVMLRSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Acute laryngeal dystonia due to clebopride simulating allergic reaction].
Mercè Corominas Sánchez et al.
Medicina clinica, 129(6), 238-239 (2007-08-07)
Domenico Bosco et al.
Clinical neuropharmacology, 32(2), 107-108 (2009-06-11)
Dystonic reactions produce twisting and repetitive movements or abnormal posturing. Severe dystonic reactions have been shown to occur in concert with numerous medications. This report details the case of a patient who developed hemifacial dystonia as acute side reaction from
M Blanco et al.
Journal of pharmaceutical and biomedical analysis, 15(3), 329-338 (1996-12-01)
A spectrophotometric method for the simultaneous determination of the active principle and a flavouring agent in syrups containing additional excipients is proposed. The calibration matrix must include all the variability expected in the samples and this is achieved using laboratory-made
E M Byrnes et al.
Developmental psychobiology, 29(8), 653-666 (1996-12-01)
D1- and D2-like antagonist-induced catalepsy and dorsal immobility were studied in pups (Day 10) and weanlings (Days 20, 28, or 35) that received intraventricular injection of 6-OHDA (50 micrograms/hemisphere) or its vehicle solution or postnatal Day 3. The ability of
I S Stovolosov et al.
Zhurnal vysshei nervnoi deiatelnosti imeni I P Pavlova, 60(2), 247-254 (2010-05-18)
A new method of studying "child's" (maternal bonding) behavior of newborn rats was developed. The efficiency of the method was proved in estimation of dopaminergic control of the infant-mother attachment. Selective D2-antagonist clebopride applied in subthreshold for motor activity doses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service