Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7784

Sigma-Aldrich

Z-Gly-Gly-Arg β-naphthylamide hydrochloride

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H33N7O5 · HCl
CAS Number:
Molecular Weight:
584.07
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

~98%

storage temp.

−20°C

SMILES string

Cl.NC(=N)NCCCC(NC(=O)CNC(=O)CNC(=O)OCc1ccccc1)C(=O)Nc2ccc3ccccc3c2

InChI

1S/C28H33N7O5.ClH/c29-27(30)31-14-6-11-23(26(38)34-22-13-12-20-9-4-5-10-21(20)15-22)35-25(37)17-32-24(36)16-33-28(39)40-18-19-7-2-1-3-8-19;/h1-5,7-10,12-13,15,23H,6,11,14,16-18H2,(H,32,36)(H,33,39)(H,34,38)(H,35,37)(H4,29,30,31);1H

InChI key

BVFJVGPYUPGGDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

A sensitive and specific fluorogenic substrate for the assay of trypsin in human serum.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kramer, S., et al.
The Journal of Surgical Research, 8, 253-253 (1968)
Jorge Frias et al.
Journal of microbiology and biotechnology, 31(2), 327-337 (2020-11-06)
Fibrinolytic enzymes with a direct mechanism of action and safer properties are currently requested for thrombolytic therapy. This paper reports on a new enzyme capable of degrading blood clots directly without impairing blood coagulation. This enzyme is also non-cytotoxic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service