Skip to Content
MilliporeSigma
All Photos(1)

Documents

C7493

Sigma-Aldrich

Aminopurinyl pyranobenzopyranone monohydrate

≥98% (HPLC), solid

Synonym(s):

(5aS,7S)-7-[2-(6-Amino-3H-purin-3-yl)-1-methylethyl]-5a,6,8,9-tetrahydro-3-methyl-1H,7H-pyrano[4,3-b]benzopyran-1-one monohydrate, CP2 monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23N5O3 · H2O
CAS Number:
Molecular Weight:
411.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.C[C@H](Cn1cnc(N)c2ncnc12)[C@H]3CCC4=CC5=C(O[C@H]4C3)C=C(C)OC5=O

InChI

1S/C21H23N5O3.H2O/c1-11(8-26-10-25-19(22)18-20(26)24-9-23-18)13-3-4-14-6-15-17(29-16(14)7-13)5-12(2)28-21(15)27;/h5-6,9-11,13,16H,3-4,7-8,22H2,1-2H3;1H2/t11?,13-,16-;/m0./s1

InChI key

WIRCSRUVUSUQDQ-HTIJIITDSA-N

Biochem/physiol Actions

Aminopurinyl pyranobenzopyranone monohydrate is an inhibitor of Aβ oligomer-induced cytotoxicity. It is a new Alzheimer′s research tool and potential therapeutic agent. Aminopurinyl pyranobenzopyranone monohydrate acts by inhibiting growth of Aβ oligomers and/or by preventing their toxic conformation or interaction with cellular substrates.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service