Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C7136

Sigma-Aldrich

Coproporphyrin I tetraethyl ester

from bovine porphyric feces

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H54N4O8
CAS Number:
Molecular Weight:
766.92
MDL number:
UNSPSC Code:
12171500

biological source

bovine porphyric feces

ε (extinction coefficient)

1,800-2,300 at 400 nm in chloroform at 1% (actual value given on label)

storage temp.

−20°C

SMILES string

CCOC(=O)CCc1c(C)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(C)c5CCC(=O)OCC)c(C)c4CCC(=O)OCC)c(C)c3CCC(=O)OCC

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service