Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6770

Sigma-Aldrich

Carbadox

Synonym(s):

3-(2-Quinoxalinylmethylene)carbazic acid methyl ester N,N′-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N4O4
CAS Number:
Molecular Weight:
262.22
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.85

form

powder

solubility

1 M NaOH: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

COC(=O)N\N=C\c1cn(=O)c2ccccc2n1=O

InChI

1S/C11H10N4O4/c1-19-11(16)13-12-6-8-7-14(17)9-4-2-3-5-10(9)15(8)18/h2-7H,1H3,(H,13,16)/b12-6+

InChI key

OVGGLBAWFMIPPY-WUXMJOGZSA-N

Application

Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities.

Biochem/physiol Actions

Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Awais Ihsan et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 51, 330-336 (2012-10-16)
Quinoxaline-1,4-dioxides (QdNOs) are the potent heterocyclic N-oxides with interesting biological properties such as antibacterial, anticandida, antitubercular, anticancer and antiprotozoal activities. Here, we tested and compared the mequindox (MEQ) for mutagenic abilities in a battery of different short term tests according
G Gizzi et al.
Food additives and contaminants, 24(11), 1226-1235 (2007-09-14)
The performance characteristics of an analytical method based on high-performance liquid chromatography (HPLC) for the detection of the banned growth promoters, carbadox and olaquindox, in feedstuff were determined via a collaborative study. The relative standard deviation of repeatability (RSDr) ranged
Qian Chen et al.
Mutation research, 638(1-2), 11-16 (2007-09-28)
Carbadox, a quinoxaline 1,4-dioxide derivative, is a known mutagen with its functional mechanism yet to be well defined. In the present study we used a shuttle vector assay in vitro to uncover the functional details of carbadox-induced mutagenesis in mammalian
L Beutin et al.
Antimicrobial agents and chemotherapy, 20(3), 336-343 (1981-09-01)
The quinoxaline-di-N-oxides carbadox, olaquindox, and quindoxin, which are potent antibacterial agents, were tested for mutagenicity in the Salmonella microsomal system. They all induced base pair substitutions and frameshift mutations in Salmonella, and occurred independently of the presence of a rat
Amisha D Shah et al.
Environmental science & technology, 40(23), 7228-7235 (2006-12-22)
The potential release of carbadox (CDX), a commonly used antibacterial agent in swine husbandry, into water systems is of a concern due to its carcinogenic and genotoxic effects. Until this study, the reactivity of carbadox (possessing quinoxaline N,N'-dioxide and hydrazone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service