Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6617

Sigma-Aldrich

CBIQ

≥98% (HPLC), solid

Synonym(s):

4-Chlorobenzo[f]isoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8NCl
CAS Number:
Molecular Weight:
213.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: 15 mg/mL

SMILES string

Clc1nccc2c1ccc3ccccc23

InChI

1S/C13H8ClN/c14-13-12-6-5-9-3-1-2-4-10(9)11(12)7-8-15-13/h1-8H

InChI key

SHQLTRRYZVBEMR-UHFFFAOYSA-N

Biochem/physiol Actions

CBIQ is an activator of the cystic fibrosis transmembrane conductance regulator (CFTR) and the intermediate-conductance calcium-sensitive K+ channel (KCNN4). CBIQ is a novel tool for a new ion channel; first activator of ΔF508 CFTR, the mutant form of CFTR chloride channel present in 75% of cystic fibrosis patients. Other benzoquinolines act at the normal CFTR but not at ΔF508 CFTR. CBIQ also activates KCNN4, which hyperpolarizes airway epithelial cells to promote anion flux, a further benefit in cystic fibrosis.

Features and Benefits

This compound is featured on the Chloride Channels and Potassium Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to chloride channels for your research needs.

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service