Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6422

Sigma-Aldrich

(±)-Cotinine-(methyl-d3)

Synonym(s):

1-(Methyl-d3)-5-(3-pyridinyl)-2-pyrrolidinone, Deuterated (±)-cotinine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D3H9N2O
CAS Number:
Molecular Weight:
179.23
MDL number:
UNSPSC Code:
12142207
PubChem Substance ID:

isotopic purity

98 atom % D

bp

250 °C/150 mmHg (lit.)

mp

40-42 °C (lit.)

mass shift

M+3

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])N1C(CCC1=O)c2cccnc2

InChI

1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/i1D3

InChI key

UIKROCXWUNQSPJ-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Major metabolite of nicotine in humans

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thompson, J.A., et al.
Journal of Chromatographic Science, 231, 53-53 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service