Skip to Content
MilliporeSigma
All Photos(2)

Documents

C6399

Sigma-Aldrich

Cholesteryl n-valerate

≥90% (HPLC; detection at 205 nm)

Synonym(s):

3β-Hydroxy-5-cholestene 3-pentanoate, 5-Cholesten-3β-ol 3-pentanoate, Cholesteryl pentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H54O2
CAS Number:
Molecular Weight:
470.77
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥90% (HPLC; detection at 205 nm)

form

solid

SMILES string

CCCCC(O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](CCCC(C)C)C)=O

InChI

1S/C32H54O2/c1-7-8-12-30(33)34-25-17-19-31(5)24(21-25)13-14-26-28-16-15-27(23(4)11-9-10-22(2)3)32(28,6)20-18-29(26)31/h13,22-23,25-29H,7-12,14-21H2,1-6H3/t23-,25?,26?,27?,28?,29?,31?,32?/m1/s1

InChI key

RWTQCZGAMKTBRV-ICUBIBKNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Use of liquid-crystal cutaneous thermography for the diagnosis of bone diseases].
I G Gertsen et al.
Ortopediia travmatologiia i protezirovanie, (2)(2), 66-69 (1981-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service