Skip to Content
MilliporeSigma
All Photos(1)

Documents

C6380

Sigma-Aldrich

Cyclohexylamine

≥98%

Synonym(s):

Aminocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NH2
CAS Number:
Molecular Weight:
99.17
Beilstein:
471175
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

3.42 (vs air)

vapor pressure

10 mmHg ( 22 °C)
23 mmHg ( 37.7 °C)

Assay

≥98%

autoignition temp.

559 °F

refractive index

n20/D 1.459 (lit.)

bp

134 °C (lit.)

mp

−17 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

NC1CCCCC1

InChI

1S/C6H13N/c7-6-4-2-1-3-5-6/h6H,1-5,7H2

InChI key

PAFZNILMFXTMIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service