Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C5051

Sigma-Aldrich

C-Peptide human

≥95% (HPLC)

Synonym(s):

Arg-Arg-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-Lys-Arg, Insulin C chain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C153H259N49O52
CAS Number:
Molecular Weight:
3616.99
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(C)C)C(C)C)C(=O)NCC(=O)NCC(=O)NCC(=O)N1CCC[C@H]1C(=O)NCC(=O)N[C@@H](C)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C153H259N49O52/c1-71(2)55-94(193-134(237)91(40-48-119(222)223)189-148(251)122(78(15)16)200-135(238)86(33-41-105(156)205)179-112(212)67-175-147(250)121(77(13)14)199-136(239)88(35-43-107(158)207)188-139(242)97(58-74(7)8)194-142(245)100(61-120(224)225)197-133(236)90(39-47-118(220)221)182-124(227)80(18)177-129(232)89(38-46-117(218)219)186-130(233)84(29-23-51-167-152(162)163)183-126(229)82(155)27-22-50-166-151(160)161)128(231)172-63-110(210)169-62-109(209)170-68-115(215)201-53-25-31-103(201)145(248)174-64-111(211)176-79(17)123(226)171-65-113(213)180-101(69-203)144(247)196-99(60-76(11)12)141(244)190-92(36-44-108(159)208)149(252)202-54-26-32-104(202)146(249)198-95(56-72(3)4)137(240)178-81(19)125(228)192-96(57-73(5)6)138(241)185-85(37-45-116(216)217)127(230)173-66-114(214)181-102(70-204)143(246)195-98(59-75(9)10)140(243)187-87(34-42-106(157)206)132(235)184-83(28-20-21-49-154)131(234)191-93(150(253)254)30-24-52-168-153(164)165/h71-104,121-122,203-204H,20-70,154-155H2,1-19H3,(H2,156,205)(H2,157,206)(H2,158,207)(H2,159,208)(H,169,210)(H,170,209)(H,171,226)(H,172,231)(H,173,230)(H,174,248)(H,175,250)(H,176,211)(H,177,232)(H,178,240)(H,179,212)(H,180,213)(H,181,214)(H,182,227)(H,183,229)(H,184,235)(H,185,241)(H,186,233)(H,187,243)(H,188,242)(H,189,251)(H,190,244)(H,191,234)(H,192,228)(H,193,237)(H,194,245)(H,195,246)(H,196,247)(H,197,236)(H,198,249)(H,199,239)(H,200,238)(H,216,217)(H,218,219)(H,220,221)(H,222,223)(H,224,225)(H,253,254)(H4,160,161,166)(H4,162,163,167)(H4,164,165,168)/t79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,121-,122-/m0/s1

InChI key

JEPNOJXFZHBCTB-ZCUALFGZSA-N

Gene Information

human ... INS(3630)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

In proinsulin the insulin A and B chains are linked by C peptide. During the biosynthesis of insulin, the C peptide promotes proper protein folding and disulfide bond placement after which it is cleaved. The cleaved peptide may inhibit insulin aggregation and is stored and released from the β cells with insulin in a one-to-one molar ratio. C peptide is a marker for β cell function in vivo; levels are enhanced in insulin resistance or insuloma and reduced in insulin insufficiency (type I diabetes). C peptide may alleviate the vascular damage and neuropathy associated with insulin depletion.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service