Skip to Content
MilliporeSigma
All Photos(1)

Documents

C4877

Sigma-Aldrich

Z-Pro-ONp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O6
CAS Number:
Molecular Weight:
370.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

O=C(N1CCC[C@H]1C(OC2=CC=C([N+]([O-])=O)C=C2)=O)OCC3=CC=CC=C3

InChI

1S/C19H18N2O6/c22-18(27-16-10-8-15(9-11-16)21(24)25)17-7-4-12-20(17)19(23)26-13-14-5-2-1-3-6-14/h1-3,5-6,8-11,17H,4,7,12-13H2/t17-/m0/s1

InChI key

GXUFIJVKXYWCAO-KRWDZBQOSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service