Skip to Content
MilliporeSigma
All Photos(1)

Documents

C4662

Sigma-Aldrich

Carbetapentane citrate salt

Synonym(s):

1-Phenylcyclopentanecarboxylic acid 2-(2-diethylaminoethoxy)ethyl ester citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31NO3 · C6H8O7
CAS Number:
Molecular Weight:
525.59
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

solubility

H2O: soluble

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)CCOCCOC(=O)C1(CCCC1)c2ccccc2

InChI

1S/C20H31NO3.C6H8O7/c1-3-21(4-2)14-15-23-16-17-24-19(22)20(12-8-9-13-20)18-10-6-5-7-11-18;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-7,10-11H,3-4,8-9,12-17H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

AKJDEXBCRLOVTH-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

High affinity σ1 agonist; ORL1 antagonist; antitussive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Shah et al.
British journal of pharmacology, 125(3), 510-516 (1998-11-07)
1. Electrical field stimulation (EFS) of guinea-pig isolated main bronchi induced a non-adrenergic non-cholinergic (NANC) contractile response. Nociceptin (0.01-1 microm) significantly inhibited the contractile response to EFS (P<0.01), but not to capsaicin (P>0.05). 2. The mu-, delta- and kappa-opioid receptor
T Kobayashi et al.
British journal of pharmacology, 120(6), 986-987 (1997-03-01)
Taking advantage of the functional coupling of the nociceptin/orphanin FQ receptor with the G-protein-activated inwardly rectifying K+ (GIRK) channel, we investigated the effects of various sigma ligands on the nociceptin/orphanin FQ receptor in Xenopus oocytes co-injected with the cloned nociceptin/orphanin
D R Wallace et al.
Journal of neurochemistry, 64(2), 700-710 (1995-02-01)
Cross-reactions between dopamine D3 and sigma receptor ligands were investigated using (+/-)-7-hydroxy-N,N-di-n-[3H]propyl-2-aminotetralin [(+/-)-7-OH-[3H]-DPAT], a putative D3-selective radioligand, in conjunction with the unlabeled sigma ligands 1,3-di(2-tolyl)guanidine (DTG), carbetapentane, and R(-)-N-(3-phenyl-1-propyl)-1-phenyl-2-aminopropane [R(-)-PPAP]. In transfected CCL1.3 mouse fibroblasts expressing the human D3 receptor
A proposal for the classification of sigma binding sites.
R Quirion et al.
Trends in pharmacological sciences, 13(3), 85-86 (1992-03-01)
S J Annels et al.
Brain research, 564(2), 341-343 (1991-11-15)
The non-opioid antitussive drugs, dextromethorphan, caramiphen and carbetapentane, are also anticonvulsant. The effects of these antitussives on potassium-stimulated release of endogenous amino acids from rabbit hippocampal slices was tested. All 3 drugs significantly reduced the release of glutamate, with carbetapentane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service