Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C4147

Sigma-Aldrich

γ-Carboxy-L-glutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO6
CAS Number:
Molecular Weight:
191.14
Beilstein:
2417114
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

N[C@@H](CC(C(O)=O)C(O)=O)C(O)=O

InChI

1S/C6H9NO6/c7-3(6(12)13)1-2(4(8)9)5(10)11/h2-3H,1,7H2,(H,8,9)(H,10,11)(H,12,13)/t3-/m0/s1

InChI key

UHBYWPGGCSDKFX-VKHMYHEASA-N

Gene Information

rat ... Grm2(24415)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gamma-carboxy-L-glutamic acid (GLA) is used as a metabotropic glutamate receptor (mGluR) subtype ligand wherein GLA is a group-II mGlu2 receptor and group-III mGlu7, mGlu8 antagonist and group-III mGlu4, mGlu6 agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Randal X Moldrich et al.
European journal of pharmacology, 476(1-2), 3-16 (2003-09-13)
Metabotropic glutamate (mGlu) receptors have multiple actions on neuronal excitability through G-protein-linked modifications of enzymes and ion channels. They act presynaptically to modify glutamatergic and gamma-aminobutyric acid (GABA)-ergic transmission and can contribute to long-term changes in synaptic function. The recent
I Brabet et al.
Neuropharmacology, 37(8), 1043-1051 (1998-12-02)
In a previous study we reported that the addition of a carboxylic group to the mGlu receptor agonist aminocyclopentane-1,3-dicarboxylate (ACPD) changes its properties from agonist to antagonist at both mGlu1 and mGlu2 receptors, and resulted in an increase in affinity
Hilde Lavreysen et al.
Current medicinal chemistry, 15(7), 671-684 (2008-03-14)
Metabotropic glutamate (mGlu) receptors have received much attention, driven by a strong belief in the potential of these modulatory glutamate receptors as drug targets. So far, major drug discovery programs have largely focused on group I (mGlu1 and 5) and
G Gerber et al.
Neuroscience, 100(2), 393-406 (2000-09-29)
The effects of group II and group III metabotropic glutamate receptor agonists on synaptic responses evoked by primary afferent stimulation in the dorsal horn, but mostly substantia gelatinosa, neurons were studied in the spinal cord slice preparation using conventional intracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service