Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-41002

Sigma-Aldrich

Trypsin / EDTA 2

0.025% Trypsin / 0.01% EDTA, 30 ml

Sign Into View Organizational & Contract Pricing

Select a Size

30 ML
$36.60

$36.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
30 ML
$36.60

About This Item

UNSPSC Code:
12352207
NACRES:
NA.78

$36.60


Please contact Customer Service for Availability

Request a Bulk Order

form

aqueous solution

packaging

pkg of 30 ml

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Trypsin / EDTA 2 (0.025% / 0.01%)

Application

Trypsin is still the most widely employed enzyme used for cell detachment. PromoCell´s Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This Trypsin/EDTA Solution has a ratio of 0.025% trypsin and 0.01% EDTA and is available in different sizes.

Quality

All lots of PromoCell Cell Detachment Reagents are subjected to comprehensive quality control tests. Each lot is routinely tested for biological function, absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria).

Storage and Stability

Store at -20°C in the dark immediately after arrival. Refrigerate thawed solution at 4 to 8°C. If stored properly, the product is stable until the expiry date stated on the label.

Subculture Routine

Click here for more information.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service