Skip to Content
MilliporeSigma
All Photos(1)

Documents

C4022

Sigma-Aldrich

DL-Cysteine

≥98% (TLC)

Synonym(s):

(±)-2-Amino-3-mercaptopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
121.16
Beilstein:
1721406
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (TLC)

mp

225 °C (dec.) (lit.)

SMILES string

NC(CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)

InChI key

XUJNEKJLAYXESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

NMDA agonist that is an agonist at quisqualate receptors at high concentration.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service