Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-39615

Sigma-Aldrich

Osteoblast Growth Medium

SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml

Synonym(s):

Recommended with C-27010

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$336.00

$336.00


Please contact Customer Service for Availability


Select a Size

Change View
1 VIAL
$336.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

$336.00


Please contact Customer Service for Availability

form

aqueous solution

packaging

pkg of 1 vial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Osteoblast Growth Medium SupplementMix (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-27010

Application

Osteoblast Growth Medium has been optimized for the in vitro cultivation of human osteoblasts. The fetal calf serum in the medium is strictly pretested for optimal performance. The medium is optimized for primary human cells, but can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.Recommended for:Human Osteoblasts (HOB)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixSupplementMix: Contains all media supplements premixed in one vial

Quality

All lots of PromoCell Osteoblast Growth Medium are subjected to comprehensive quality control tests using primary human osteoblasts. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested osteoblasts. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.1 ml / ml

Storage and Stability

Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human osteoblasts and media for the optimal growth and mineralization of human osteoblast cultures. Protocols for handling, cell passaging, media, and product use.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service