Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-39170

Sigma-Aldrich

Small Airway Epithelial Cell Growth Medium

SupplementPack containing all media supplements as individual vials, 1 Pack for 500 ml

Synonym(s):

Recommended with C-21270

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

packaging

pkg of 1 kit

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Small Airway Epithelial Cell Growth Medium SupplementPack (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-21270

Application

Small Airway Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the distal respiratory system. The optimal adjustment of growth factors in this medium ensures excellent in vitro growth. The medium is optimized for primary human cells but can also be used for porcine small airway epithelial cells.Recommended for:Human Small Airway Epithelial Cells (HSAEpC)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Small Airway Epithelial Cell Growth Medium are subjected to comprehensive quality control tests using primary human small airway epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested small airway epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / mlEpidermal Growth Factor (recombinant human): 10 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.5 µg / mlEpinephrine: 0.5 µg / mlTriiodo-L-thyronine: 6.7 ng / mlTransferrin (recombinant human): 10 µg / mlRetinoic Acid: 0.1 ng / mlBovine Serum Albumin-Fatty Acid Free (BSA-FAF): 2.5 mg/ml

Storage and Stability

Store the SupplementPack at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service