Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3255

Sigma-Aldrich

Corynanthine

Synonym(s):

17α-Hydroxyyohimban-16β-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O3
CAS Number:
Molecular Weight:
354.44
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@H](O)[C@@H](C(=O)OC)[C@@]1([H])C[C@]3([H])N(CCc4c3[nH]c5ccccc45)C2

InChI

1S/C21H26N2O3/c1-26-21(25)19-15-10-17-20-14(13-4-2-3-5-16(13)22-20)8-9-23(17)11-12(15)6-7-18(19)24/h2-5,12,15,17-19,22,24H,6-11H2,1H3/t12-,15-,17-,18-,19-/m0/s1

InChI key

BLGXFZZNTVWLAY-DKJBZYCGSA-N

Gene Information

human ... CYP2D6(1565)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei Chen et al.
Journal of AOAC International, 98(4), 896-901 (2015-08-14)
A single-laboratory validation was performed on a practical ultra-HPLC (UHPLC)-diode array detector (DAD)/tandem MS method for determination of yohimbine in yohimbe barks and related dietary supplements. Good separation was achieved using a Waters Acquity ethylene bridged hybrid C18 column with
Fernando Huete-Toral et al.
The Journal of pharmacology and experimental therapeutics, 352(1), 119-128 (2014-10-26)
Melatonin and its analog 5-MCA-NAT (5-methylcarboxyamino-N-acetyl tryptamine) are active compounds reducing intraocular pressure (IOP). This action is mediated through MT2 and the putative MT3 melatonin receptor, producing a transient reduction of IOP that lasts for a few hours and has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service