Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3115

Sigma-Aldrich

Control Primer Set for Arabidopsis CBP20 Gene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

storage temp.

−20°C

General description

This product consists of a primer set for the Arabidopsis housekeeping gene CBP20 (At5g44200). CBP20 encodes a nuclear cap-binding protein. It is intended for use as an internal control for Q-RT-PCR for expression analysis of any Arabidopsis gene.

Of our three control primer series for Arabidopsis gene expression, the CBP20 control primer is designed as a control for medium- to low-abundant gene expression. The other two control primers in the series are ubiquitin (UBC, Product Code C3240) also designed as a control for medium- to low-abundant gene expression and actin-2 (Product Code C3615) for high-abundant gene expression.

Components

Each tube contains enough primer (forward or reverse) for 100 RT-PCR reactions (50 μl reaction volume)
  • Forward Primer (Tm = 68 °C based on % GC method in 1 M salt), 20 μM, in TE buffer, Product Code C2740.
  • Reverse Primer (Tm = 66 °C based on % GC method in 1 M salt), 20 μM, in TE buffer, Product Code C2865.

Suitability

Primer should be stored at -20 °C. Primers are stable at -20 °C for at least 1 year and can be frozen and thawed at least 10 times.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatjana Trcek et al.
Molecular cell, 78(5), 941-950 (2020-05-29)
mRNAs enriched in membraneless condensates provide functional compartmentalization within cells. The mechanisms that recruit transcripts to condensates are under intense study; however, how mRNAs organize once they reach a granule remains poorly understood. Here, we report on a self-sorting mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service