Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C3026

Sigma-Aldrich

Colicin E1 from Escherichia coli

lyophilized powder, 10,000-20,000 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

specific activity

10,000-20,000 units/mg protein

composition

Protein, ~30% Lowry

storage temp.

2-8°C

Packaging

Package size based on protein content

Unit Definition

One unit per mL is the minimal concentration required to cause a zone of clearing on a lawn of E. coli ATCC9637 cells.

Preparation Note

Prepared by a modification of the method of Schwartz, S. and Helinski, D., J. Biol. Chem., 246, 6318 (1971).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service