Skip to Content
MilliporeSigma
All Photos(1)

Documents

C2859

Sigma-Aldrich

6-Carboxytetramethylrhodamine

Synonym(s):

6-TAMRA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22N2O5
CAS Number:
Molecular Weight:
430.45
MDL number:
UNSPSC Code:
12171500

description

single isomer

SMILES string

CN(C)c1ccc2c(OC3=C\C(C=CC3=C2c4cc(ccc4C([O-])=O)C(O)=O)=[N+](\C)C)c1

InChI

1S/C25H22N2O5/c1-26(2)15-6-9-18-21(12-15)32-22-13-16(27(3)4)7-10-19(22)23(18)20-11-14(24(28)29)5-8-17(20)25(30)31/h5-13H,1-4H3,(H-,28,29,30,31)

InChI key

COCMHKNAGZHBDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The corresponding succinimidyl ester is a single isomer fluorophore most commonly used in automated DNA sequencing.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service