Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C2470

Sigma-Aldrich

Coenzyme Q4

≥90%

Synonym(s):

2,3-Dimethoxy-5-methyl-6-(geranylgeranyl)-1,4-benzoquinone, Q-4, Ubiquinone-20, Ubiquinone-4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42O4
CAS Number:
Molecular Weight:
454.64
Beilstein:
2066531
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.51

Assay

≥90%

form

liquid

storage temp.

−20°C

SMILES string

COC1C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C)=C(C)C1=O

InChI

1S/C29H44O4/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-17-23(5)18-19-25-24(6)26(30)28(32-7)29(33-8)27(25)31/h12,14,16,18,28-29H,9-11,13,15,17,19H2,1-8H3/b21-14+,22-16+,23-18+

InChI key

MDLQEORKYVBTDV-INVBOZNNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coenzyme Q4 (CoQ4) is a 4 isoprenyl group member of a family of ubiquinones that share a quinine chemical group but differ in the number of isoprenyl chemical subunits in their tail. The CoQ compounds are lipid soluble components of cell membranes where they perform multiple functions such as electron and proton transport. The most well studied CoQ compound is CoQ10. CoQ4 is frequently used in comparison studies on the effect of isoprenyl chain length on CoQ functions or distribution.

Biochem/physiol Actions

Increases the fluorescence polarization of perylene incorporated into ubiquinone-depleted mitochondrial and bilayer vesicles prepared from mitochondrial phospholipids

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolanta Dłużewska et al.
PloS one, 11(7), e0159629-e0159629 (2016-07-28)
In the present studies, we focused on substrate specificity of tocopherol cyclase, the key enzyme in the biosynthesis of the tocopherols and plastochromanol-8, the main plant lipid antioxidants, with special emphasis on the preference for tocopherols and plastochromanol-8 precursors, taking
Italo José B Durval et al.
Marine pollution bulletin, 157, 111357-111357 (2020-07-14)
In this study, Bacillus cereus was cultivated in a mineral medium composed of 2% frying oil and 0.12% peptone to produce a biosurfactant. The production was scaled up from flasks to 1.2-, 3.0- and 50-L bioreactors, where surface tension achieved
V E Kagan et al.
Free radical biology & medicine, 9(2), 117-126 (1990-01-01)
Ubiquinones (CoQn) are intrinsic lipid components of many membranes. Besides their role in electron-transfer reactions they may act as free radical scavengers, yet their antioxidant function has received relatively little study. The efficiency of ubiquinols of varying isoprenoid chain length
T Kishi et al.
Proceedings of the National Academy of Sciences of the United States of America, 73(12), 4653-4656 (1976-12-01)
Adriamycin inhibits the succinoxidase system and the NADH-oxidase system. Both of the intact mitocondrial enzymes and the pentane-extracted preparations are inhibited. The inhibition can be prevented by a molar ratio of coenzyme to adriamycin of 3:1 for coenzyme Q10 (ubiquinone)
Zhiliang Hei et al.
Molecules (Basel, Switzerland), 25(1) (2020-01-08)
Gadus morhua eggs contain several nutrients, including polyunsaturated fatty acids, lecithin and glycoproteins. A novel sialoglycopeptide from the eggs of G. morhua (Gm-SGPP) was extracted with 90% phenol and purified by Q Sepharose Fast Flow (QFF) ion exchange chromatography, followed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service