Skip to Content
MilliporeSigma
All Photos(1)

Documents

C2353

Supelco

Chromatography columns, general-purpose

volume 13 mL, Overall H 7 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

material

polypropylene

technique(s)

LPLC: suitable

Overall H

7 in.

volume

13 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Precision molded from premium-grade, chemically inert plastic for organic solvents
  • Includes reservoir & end caps
  • Polyethylene filter matrix support in medium porosity

Application

For use in general chromatographic work for extraction of components, with special applications in isoenzyme and protein studies, hemoglobin fractionation and desalting.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service