Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C2253

Sigma-Aldrich

N-Chloroacetyl-L-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12ClNO3
CAS Number:
Molecular Weight:
193.63
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

form

solid

technique(s)

ligand binding assay: suitable

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)CCl)C(O)=O

InChI

1S/C7H12ClNO3/c1-4(2)6(7(11)12)9-5(10)3-8/h4,6H,3H2,1-2H3,(H,9,10)(H,11,12)/t6-/m0/s1

InChI key

LJRISAYPKJORFZ-LURJTMIESA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Tokuyama et al.
Applied microbiology and biotechnology, 42(6), 853-859 (1995-03-01)
Thermostable N-acylamino acid racemase from Amycolatopsis sp. TS-1-60, a rare actinomycete strain selected for its ability to grow on agar plates incubated at 40 degrees C, was purified to homogeneity and characterized. The relative molecular mass (M(r)) of the native

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service