Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C213130

Sigma-Aldrich

DMT-2′O-Methyl-rC(ac) Phosphoramidite

configured for ABI

Synonym(s):

N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H52N5O9P
CAS Number:
Molecular Weight:
801.86
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

non-animal source (no BSE/TSE risk)

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.1% single unspecified Impurity (reversed phase HPLC)
≤0.3% mC2 (reversed phase HPLC, Hydrolysate)
≤0.3% mC3 (reversed phase HPLC, DMT-rC(ac)me)
≤0.3% mC4 (reversed phase HPLC, DMT-rC(ac)me-Ac)
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤1.0% mC1 (reversed phase HPLC, DMT-rC(ac)me-DMT)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for ABI

nucleoside profile

base: cytidine
base protecting group: acetyl
2' protecting group: methyl
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

P(N(C(C)C)C(C)C)(O[C@@H]1[C@H](O[C@H](C1OC)N5C=CC(=NC5=O)NC(=O)C)COC(c4ccc(cc4)OC)(c3ccc(cc3)OC)c2ccccc2)OCCC#N

InChI

1S/C42H52N5O9P/c1-28(2)47(29(3)4)57(54-26-12-24-43)56-38-36(55-40(39(38)52-8)46-25-23-37(44-30(5)48)45-41(46)49)27-53-42(31-13-10-9-11-14-31,32-15-19-34(50-6)20-16-32)33-17-21-35(51-7)22-18-33/h9-11,13-23,25,28-29,36,38-40H,12,26-27H2,1-8H3,(H,44,45,48,49)/t36-,38-,39?,40-,57?/m1/s1

InChI key

WNWUMIPFLOKTEZ-IXZONTHDSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service