Skip to Content
MilliporeSigma
All Photos(1)

Documents

C-21275

Sigma-Aldrich

Small Airway Epithelial Cell Growth Medium

Basal Medium, phenol red-free, 500 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

aqueous solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Small Airway Epithelial Cell Basal Medium, phenol red-free (supplements sold separately)

Application

Small Airway Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the distal respiratory system. The optimal adjustment of growth factors in this medium ensures excellent in vitro growth. The medium is optimized for primary human cells but can also be used for porcine small airway epithelial cells.Recommended for:Human Small Airway Epithelial Cells (HSAEpC)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Small Airway Epithelial Cell Growth Medium are subjected to comprehensive quality control tests using primary human small airway epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested small airway epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service