Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C2051

Sigma-Aldrich

[Asn-OH17]-Conantokin G

≥97% (HPLC)

Synonym(s):

Conantokin-G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C88H138N26O44
CAS Number:
Molecular Weight:
2264.19
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Amino Acid Sequence

Gly-Glu-γ-CarboxyGlu-γ-CarboxyGlu-Leu-Gln-γ-CarboxyGlu-Asn-Gln-γ-CarboxyGlu-Leu-Ile-Arg-γ-CarboxyGlu-Lys-Ser-Asn

Linkage

Carboxy-terminal free acid analog of sleep-inducing peptide conantokin-G

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Haack et al.
The Journal of biological chemistry, 265(11), 6025-6029 (1990-04-15)
Conantokin-T, a 21-amino acid peptide which induces sleep-like symptoms in young mice was purified from the venom of the fish-hunting cone snail, Conus tulipa. The amino acid sequence of the peptide was determined and verified by chemical synthesis. The peptide
B M Olivera et al.
Science (New York, N.Y.), 249(4966), 257-263 (1990-07-20)
Conus venoms contain a remarkable diversity of pharmacologically active small peptides. Their targets are ion channels and receptors in the neuromuscular system. The venom of Conus geographus contains high-affinity peptides that act on voltage-sensitive calcium channels, sodium channels, N-methyl-D-aspartate (NMDA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service