Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C1791

Sigma-Aldrich

Cresyl Violet acetate

Technical grade.

Synonym(s):

9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N3O3
CAS Number:
Molecular Weight:
321.33
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

technical grade

composition

Dye content, ~70%

mp

140-143 °C (lit.)

SMILES string

CC(O)=O.Nc1cc2OC3=CC(=N)C=CC3=Nc2c4ccccc14

InChI

1S/C16H11N3O.C2H4O2/c17-9-5-6-13-14(7-9)20-15-8-12(18)10-3-1-2-4-11(10)16(15)19-13;1-2(3)4/h1-8,17H,18H2;1H3,(H,3,4)

InChI key

ORIKHDNZZCTIAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Recommended substitute for Cresyl echt Violet (Cresyl Fast Violet)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Ting Liu et al.
Behavioural brain research, 210(1), 24-31 (2010-02-02)
Considerable evidence has emerged supporting the neuroprotective and cognition-preserving effects of estrogen, but these benefits are complicated by the evidence that estrogen increases the risk of certain cancers. Selective estrogen receptor modulators (SERMs) that specifically target the brain while avoiding
Estelle Rousselet et al.
Journal of visualized experiments : JoVE, (69)(69), doi:10-doi:10 (2012-11-22)
Stroke is the third cause of mortality and the leading cause of disability in the World. Ischemic stroke accounts for approximately 80% of all strokes. However, the thrombolytic tissue plasminogen activator (tPA) is the only treatment of acute ischemic stroke
Priscilla Hirst et al.
Neural plasticity, 2012, 197264-197264 (2012-07-14)
Recent studies suggest that exposure to only one component of audiovisual events can lead to cross-modal cortical activation. However, it is not certain whether such crossmodal recruitment can occur in the absence of explicit conditioning, semantic factors, or long-term associations.
Sung In Cho et al.
Bioelectromagnetics, 33(7), 568-574 (2012-04-13)
Our previous study has shown that an extremely low-frequency magnetic field (ELF-MF) induces nitric oxide (NO) synthesis by Ca(2+) -dependent NO synthase (NOS) in rat brain. The present study was designed to confirm that ELF-MF affects neuronal NOS (nNOS) in
Simon C Spanswick et al.
PloS one, 7(8), e43698-e43698 (2012-08-29)
Recent evidence suggests that the medial prefrontal cortex (MFC) is important for processing contextual information. Here we evaluate the performance of mice with MFC damage in a discrimination task that requires an association between an object and the context in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service