Skip to Content
MilliporeSigma
All Photos(1)

Documents

C1538

Sigma-Aldrich

Complement C8 deficient serum human

for complement assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

form

solution

origin

USA origin

technique(s)

activity assay: suitable

impurities

infectious agent, tested

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C8A(731)

Application

Complement C8 is one of the end terminals of the complement system contained in the membrane attack complex (MAC). A deficiency of C8 may result in an increased susceptibility to Neisseria meningitidis. The most common mutation resulting in a C8 deficiency is a C to T transition in exon 9 of the C8 beta gene. Recent research however, has also discovered that two separate heterogeneous mutations may result in C8 deficiency in patients displaying recurrent meningococcal meningitis. These mutations are a duplication mutation on exon 7 and a mutation on exon 3.

Biochem/physiol Actions

Serum naturally deficient in C8 may actually contain some C8, though in greatly reduced quantity. Terminal complement complex is present at trace levels in such sera.

Physical form

Supplied as a solution in PBS, pH 7.3

Analysis Note

C8 is depleted by immunoadsorption as judged by a highly sensitive hemolytic assay.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T E Mollnes et al.
Scandinavian journal of immunology, 24(3), 307-312 (1986-09-01)
The terminal complement complex (TCC) was quantified in sera from patients with a genetic deficiency of C8 alpha-gamma or C8 beta. The individual sera contained only trace amounts of TCC compared with a normal serum pool. The content of TCC
L Saucedo et al.
Journal of immunology (Baltimore, Md. : 1950), 155(10), 5022-5028 (1995-11-15)
We studied the molecular bases for C8 beta deficiency in 34 unrelated families from the United States and the former Soviet Union. These families represented 69 unrelated null alleles of which 59 (86%) were found to be due to a
W P Kolb et al.
Journal of immunology (Baltimore, Md. : 1950), 122(5), 2103-2111 (1979-05-01)
C1q, a subcomponent of the first component of complement, has been isolated from human serum in fully hemolytically active form by affinity column chromatography and gel filtration with Bio-Gel A-5M. The affinity column was prepared by covalent coupling of purified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service