Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-12580

Sigma-Aldrich

Human Skeletal Muscle Cells (SkMC)

500,000 proliferating cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$2,460.00

$2,460.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$2,460.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$2,460.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human skeletal muscle

packaging

pkg of 500,000 cells

morphology

(smooth muscle)

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

room temp

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Cell Line Origin

Muscle

Application

Primary Human Skeletal Muscle Cells (SkMC) are isolated from different skeletal muscles (e.g. Musculus pectoralis major) from adult single donors (lot specific source information is available on request) and are provided in a proliferating format. They are positive for sarcomeric myosin and negative for smooth muscle specific α-actin.New skeletal muscle cells originate from quiescent satellite cells, which are located in the muscle fibers between the basal lamina and the sarcolemma. Quiescent satellite cells are activated by stimuli such as muscle damage. After activation, the cells, now called myoblasts, start to proliferate and fuse with damaged muscle fibers or with one another forming new myotubes.SkMC are optimal for in vitro muscle studies. They proliferate very well in the mitogen-rich PromoCell Skeletal Muscle Cell Growth Medium. Fusion to myotubes with typical multinucleated syncytia can be induced by using the PromoCell Skeletal Muscle Cell Differentiation Medium.

Components

Mitogen-rich PromoCell Skeletal Muscle Cell Growth Medium

Quality

Rigid quality control tests are performed for each lot of Human Skeletal Muscle Cells. They are tested for cell morphology, adherence rate, and cell viability. Immunohistochemical tests for cell-type specific markers are carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. Furthermore, the capacity to differentiate into multinucleated syncytia is routinely checked for each lot. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 3500 - 7000 cells per cm2Passage After Thawing: P2Tested Markers: Sarcomeric myosin positiveDifferentiation capacity to multinucleate syncytia testedGuaranteed population doublings: >15

Recommended products

Recommended Primary Cell Culture Media:Link Link

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human skeletal muscle cells and culture medium optimized for the in vitro cultivation and differentiation of primary human skeletal muscle cells. Protocols for handling, cell passaging, media, and product use.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service