Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-12463

Sigma-Aldrich

Normal Human Epidermal Melanocytes 3 (NHEM 3)

Adult, isolated in PMA-free Melanocyte Growth Medium M3, 500,000 proliferating cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,580.00

$1,580.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$1,580.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$1,580.00


Please contact Customer Service for Availability

Request a Bulk Order

packaging

pkg of 500,000 cells

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

no temp limit

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Application

Primary Normal Human Epidermal Melanocytes (NHEM) are isolated from the epidermis of adult skin (different locations) and are provided in a proliferating format. Melanocytes are located in the stratum basale, but branch out between the keratinocytes in suprabasal layers. About 5-10 % of the cells in the epidermis are melanocytes. The main purpose of melanocytes is the production of melanin, the protein responsible for the pigmentation of the skin, eyes, and hair. Melanin protects the cells in the skin and in deeper layers from the hazardous effects of UV radiation. It is produced and stored in melanosomes, which are located close to the melanocyte membrane, but it can also be transferred to neighboring keratinocytes.
Normal Human Epidermal Melanocytes M3 are isolated using the serum-free, BPE-free and PMA-free Melanocyte Growth Medium M3. Since PMA is a tumor promoting mitogen, it can interfere with experimental approaches. Therefore, we recommend using cells isolated in Melanocyte Growth Medium M3 and also using this medium for cultivation. Normal Human Epidermal Keratinocytes (NHEK) and Normal Human Dermal Fibroblasts (NHDF) from the same donor are available on request.

Quality

Rigid quality control tests are performed for each lot of Normal Human Epidermal Melanocytes. They are tested for cell morphology, adherence rate and cell viability. Furthermore, immunohistochemical tests for the cell-type specific marker, Mel-5, are carried out for each lot. Growth performance is tested through multiple passages up to 15 population doublings (PD) under culture conditions without antibiotics and antimycotics. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Culture Medium

Melanocyte Growth Medium M3 (C-4310)

Subculture Routine

Click here for more information.

Other Notes

Recommended Plating Density: 5000 - 10000 cells per cm2Passage After Thawing: P2Tested Markers: Mel-5 positiveGuaranteed population doublings: > 15

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service